8-Bromoadenine
8-Bromoadenine
8-Bromoadenine
Thermo Scientific Chemicals

8-Bromoadenine

CAS: 6974-78-3 | C5H4BrN5 | 214.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
1 g
Catalog number J65582.MC
also known as J65582-MC
Price (USD)
85.65
Online Exclusive
95.00
Save 9.35 (10%)
Each
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
95.00
Save 9.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS6974-78-3
IUPAC Name8-bromo-7H-purin-6-amine
Molecular FormulaC5H4BrN5
InChI KeyFVXHPCVBOXMRJP-UHFFFAOYSA-N
SMILESNC1=C2NC(Br)=NC2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)95% min.
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only