5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine, 98%
5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine, 98%
5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine, 98%
Thermo Scientific Chemicals

5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine, 98%

CAS: 114745-26-5 | C36H39N5O8 | 669.74 g/mol
Have Questions?
Catalog NumberQuantity
J65583.03
also known as J65583-03
1 g
Catalog number J65583.03
also known as J65583-03
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Specifications
CAS114745-26-5
Chemical Name or Material5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine
ColorWhite to Yellow
Formula Weight669.72
IUPAC NameN-[9-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxy-3-methoxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in acetonitrile

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only