Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number J65585.22
also known as J65585-22
Price (USD)
33.40
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
33.40
Each
Chemical Identifiers
CAS60-00-4
IUPAC Name2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular FormulaC10H16N2O8
InChI KeyKCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILESOC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (unspecified)99.4-100.6%
Residue on ignition0.2% max.
Impurity contentInsoluble in dilute ammonium hydroxide: 0.005% max
Impurity contentIron: 0.005% max.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only