Water, Endotoxin-free
Water, Endotoxin-free
Water, Endotoxin-free
Water, Endotoxin-free
Thermo Scientific Chemicals

Water, Endotoxin-free

CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J65589.K2
also known as J65589-K2
Price (USD)
89.65
Online Exclusive
99.10
Save 9.45 (10%)
Each
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
89.65
Online Exclusive
99.10
Save 9.45 (10%)
Each
Add to cart
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Endotoxin≤0.005 EU / mL
Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.

Solubility
Miscible with methanol, acetone and ethanol.
RUO – Research Use Only

General References:

  1. Narayanan, D.; Geena, M. G.; Lakshmi, H.; Koyakutty, M.; Nair, S.; Menon, D. Poly-(ethylene glycol) modified gelatin nanoparticles for sustained delivery of the anti-inflammatory drug Ibuprofen-Sodium: An in vitro and in vivo analysis. Medicine 2013, 9 (6), 818-828.
  2. Madhubanti, B.; Kumar, M. N.; Mrinal, S. Toll-like receptor (TLR) 4 in mrigal (Cirrhinus mrigala) : Response to lipopolysaccharide treatment and Aeromonas hydrophila infection. Int. Res. J. Biological Sci. 2013, 2 (4), 20-27.