2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%
Thermo Scientific Chemicals

2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%

C14H18FN5O5
Have Questions?
Catalog NumberQuantity
J65608.03
also known as J65608-03
1 g
Catalog number J65608.03
also known as J65608-03
Price (USD)
147.65
Special offer
Online exclusive
Ends: 30-Jun-2026
174.00
Save 26.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
147.65
Special offer
Online exclusive
Ends: 30-Jun-2026
174.00
Save 26.35 (15%)
Each
Add to cart
Specifications
Chemical Name or Material2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine
ColorWhite to Yellow
Formula Weight355.32
MDL NumberMFCD15145246
Molecular FormulaC14H18FN5O5
View more
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine is used in drugs, cosmetics or foodstuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-N2-isobutyryl-2′-deoxyguanosine is used in drugs, cosmetics or foodstuffs.

Solubility
Soluble in methanol

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only