10058-F4
10058-F4
10058-F4
Thermo Scientific Chemicals

10058-F4

CAS: 403811-55-2 | C12H11NOS2 | 249.346 g/mol
Have Questions?
Catalog NumberQuantity
J65650.MA
also known as J65650-MA
10 mg
Catalog number J65650.MA
also known as J65650-MA
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Chemical Identifiers
CAS403811-55-2
IUPAC Name5-[(4-ethylphenyl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular FormulaC12H11NOS2
InChI KeySVXDHPADAXBMFB-UHFFFAOYSA-N
SMILESCCC1=CC=C(C=C2SC(=S)NC2=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)Yellow
Purity≥98.0%
A c-Myc inhibitor that induces apoptosis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only