Dihydrofluorescein diacetate, 97%
Dihydrofluorescein diacetate, 97%
Dihydrofluorescein diacetate, 97%
Thermo Scientific Chemicals

Dihydrofluorescein diacetate, 97%

Fluorescent dye for detecting intracellular oxidants | CAS: 35340-49-9 | C24H18O7 | 418.401 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number J65759.03
also known as J65759-03
Price (USD)
201.65
Special offer
Online exclusive
Ends: 30-Jun-2026
237.00
Save 35.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
201.65
Special offer
Online exclusive
Ends: 30-Jun-2026
237.00
Save 35.35 (15%)
Each
Specifications
Beilstein364167
CAS35340-49-9
Chemical Name or MaterialDihydrofluorescein diacetate
ColorWhite to Yellow
Formula Weight418.39
View more
Fluorescent dye for detecting intracellular oxidants

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only