Cesium chloride, ultrapure, 99.9% (metals basis)
Cesium chloride, ultrapure, 99.9% (metals basis)
Cesium chloride, ultrapure, 99.9% (metals basis)
Thermo Scientific Chemicals

Cesium chloride, ultrapure, 99.9% (metals basis)

CAS: 7647-17-8 | ClCs | 168.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number J65950.14
also known as J65950-14
Price (USD)
78.65
Online Exclusive
86.70
Save 8.05 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
78.65
Online Exclusive
86.70
Save 8.05 (9%)
Each
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or crystalline powder
Total Metal Impurities≤0.1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only