This product is animal origin and can only be shipped within the United States
Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 7.0
Thermo Scientific Chemicals

Albumin, Bovine, Cohn Fraction V, 98%, Low Fatty Acid, pH 7.0

For research use
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65969.22
also known as J65969-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS9048-46-8
MDL NumberMFCD00145743
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Fatty Acid: 1mg/g max.
Assay (unspecified)Moisture: 5.0% max.
Assay (unspecified)Protease: None detected
Assay (unspecified)Viral agents: None detected
pH6.7-7.3, 1% solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only