Clarithromycin, 960-1040 ^mg/mg
Clarithromycin, 960-1040 ^mg/mg
Clarithromycin, 960-1040 ^mg/mg
Thermo Scientific Chemicals

Clarithromycin, 960-1040 ^mg/mg

CAS: 81103-11-9 | C38H69NO13 | 747.964 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66005.06
also known as J66005-06
Price (USD)
294.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
294.00
Each
Specifications
Boiling Point86°C
CAS81103-11-9
ChEBICHEBI:3732
Chemical Name or MaterialClarithromycin
Flash Point>110°C (230°F)
View more
A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only