Toluidine Blue O, Certified, 65% min dye content
Toluidine Blue O, Certified, 65% min dye content
Toluidine Blue O, Certified, 65% min dye content
Toluidine Blue O, Certified, 65% min dye content
Thermo Scientific Chemicals

Toluidine Blue O, Certified, 65% min dye content

CAS: 92-31-9 | C15H16ClN3S | 305.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
250 g
Catalog number J66015.14
also known as J66015-14
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.70
Save 8.05 (14%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.70
Save 8.05 (14%)
Each
Add to cart
Chemical Identifiers
CAS92-31-9
IUPAC Name3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
Molecular FormulaC15H16ClN3S
InChI KeyHNONEKILPDHFOL-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Dye)(as is basis): 65% min
Loss on Drying8.0% max
Solution Test(0.1% distilled water): Passes Test
Specific Absorbance (E 1%/1cm)(lambda max 650nm): >1200
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only