Tiamulin fumarate, 95%
Tiamulin fumarate, 95%
Tiamulin fumarate, 95%
Thermo Scientific™

Tiamulin fumarate, 95%

CAS: 55297-96-6 | C32H51NO8S | 609.82 g/mol
Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66020.ME
also known as J66020-ME
Price (USD)
250.65
Online Exclusive
278.00
Save 27.35 (10%)
Each
Request bulk or custom format
Price (USD)
250.65
Online Exclusive
278.00
Save 27.35 (10%)
Each
Specifications
CAS55297-96-6
EINECS Number259-581-6
Flash Point>110°(230°F)
Formula Weight609.81
IUPAC Name(2E)-but-2-enedioic acid; (2R,3S,4S,6R,8S,14R)-4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[2-(diethylamino)ethyl]sulfanyl}acetate
View more
Targets Gram-positive bacteria as well as a few mycoplasma species

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Targets Gram-positive bacteria as well as a few mycoplasma species