2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide
Thermo Scientific Chemicals

2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide

CAS: 14227-66-8 | C26H35BrO17 | 699.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number J66024.06
also known as J66024-06
Price (USD)
150.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
150.00
Each
Add to cart
Chemical Identifiers
CAS14227-66-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale to Yellow
FormSolid
Proton NMRConforms
Purity≥98.0%
It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water. Slightly soluble in ethanol

Notes
Hygroscopic, store away from water/moisture. Keep the container tightly closed. Store away from oxidizing agent. Store under dry inert gas.
RUO – Research Use Only