Specifications
CAS28244-99-7
Chemical Name or Material4-Methylphenyl 2,3,4,6-tetra-O-acetyl-beta-D-thiogalactopyranoside
ColorWhite
Formula Weight454.5
IUPAC Name[(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-methylphenyl)sulfanyloxan-2-yl]methyl acetate
View more