D-Glucuronamide, 98%
D-Glucuronamide, 98%
D-Glucuronamide, 98%
Thermo Scientific Chemicals

D-Glucuronamide, 98%

CAS: 3789-97-7 | C6H11NO6 | 193.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J66047.03
also known as J66047-03
Price (USD)
27.90
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
27.90
Each
Chemical Identifiers
CAS3789-97-7
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5%
FormPowder or crystalline powder or crystals
Appearance (Color)White to pale yellow to pale orange
Optical Rotation+32.0° to +38.0° (c=2 in water after 24hrs)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only