Apelin-12
Thermo Scientific™

Apelin-12

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J66079.LB0
also known as J66079-#0
5 mg
Catalog number J66079.LB0
also known as J66079-#0
Price (USD)
-
Quantity:
5 mg
Request bulk or custom format
Specifications
FormatLyophilized Powder
Molecular Weight (g/mol)1422.70
Product TypeApelin-12
Purification MethodHPLC
Quantity5 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C