Cefixime trihydrate
Cefixime trihydrate
Cefixime trihydrate
Thermo Scientific Chemicals

Cefixime trihydrate

A broad spectrum third generation cephalosporin | CAS: 125110-14-7 | C16H15N5O7S2 | 453.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66094.03
also known as J66094-03
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS125110-14-7
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream to pale yellow
Assay from Supplier's CofA≥97.0%
Water Content9.0-12.0%
A broad spectrum third generation cephalosporin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only