[Glp1]-Apelin-13, human, bovine
Thermo Scientific Chemicals

[Glp1]-Apelin-13, human, bovine

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66114.EXD
also known as J66114-!D
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.30
Save 13.65 (15%)
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.30
Save 13.65 (15%)
Each
Specifications
FormatLyophilized Powder
Molecular Weight (g/mol)1533.79
Product Type[Glp1]-Apelin-13, human, bovine
Purification MethodHPLC
Quantity0.5 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C