Parathyroid Hormone (1-34), human
Parathyroid Hormone (1-34), human
Parathyroid Hormone (1-34), human
Parathyroid Hormone (1-34), human
Thermo Scientific Chemicals

Parathyroid Hormone (1-34), human

Have Questions?
Catalog NumberQuantity
J66180.MCR
also known as J66180-M^
1 mg
Catalog number J66180.MCR
also known as J66180-M^
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Add to cart
Specifications
CAS52232-67-4
FormatSolid
IUPAC Name(4S)-4-{[(1S,2S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-[({[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-3-carbamoyl-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-2-carbamoyl-1-{[(1S)-1-carboxy-2-phenylethyl]carbamoyl}ethyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl]carbamoyl}-2-methylpropyl]carbamoyl}-2-carboxyethyl]carbamoyl}propyl]carbamoyl}-3-methylbutyl]carbamoyl}pentyl]carbamoyl}pentyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-3-methylbutyl]carbamoyl}-2-(1H-indol-3-yl)ethyl]carbamoyl}-3-carboxypropyl]carbamoyl}-2-methylpropyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-3-carboxypropyl]carbamoyl}-3-(methylsulfanyl)propyl]carbamoyl}-2-hydroxyethyl]carbamoyl}-2-carbamoylethyl]carbamoyl}-3-methylbutyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl]carbamoyl}pentyl]carbamoyl}methyl)carbamoyl]-3-methylbutyl]carbamoyl}-2-carbamoylethyl]carbamoyl}-2-(1H-imidazol-5-yl)ethyl]carbamoyl}-3-(methylsulfanyl)propyl]carbamoyl}-3-methylbutyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}-2-methylbutyl]carbamoyl}-4-[(2S)-2-[(2S)-2-[(2S)-2-amino-3-hydroxypropanamido]-3-methylbutanamido]-3-hydroxypropanamido]butanoic acid; acetic acid
InChI KeyBUUKFBVDKSFMHN-LKMAISLMSA-N
Molecular FormulaC183H295N55O53S2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Contents & Storage
Store at -20°C