Praziquantel, 98%
Praziquantel, 98%
Praziquantel, 98%
Thermo Scientific Chemicals

Praziquantel, 98%

Used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic. | CAS: 55268-74-1 | C19H24N2O2 | 312.413 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66188.14
also known as J66188-14
Price (USD)
277.65
Special offer
Online exclusive
Ends: 30-Jun-2026
326.00
Save 48.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
277.65
Special offer
Online exclusive
Ends: 30-Jun-2026
326.00
Save 48.35 (15%)
Each
Specifications
Beilstein761557
CAS55268-74-1
Chemical Name or MaterialPraziquantel
EINECS Number259-559-6
Flash Point>110°C (230°F)
View more
Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.

Solubility
Freely soluble in ethanol or dichloromethane. Slightly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only