Glucagon-Like Peptide-1 (7-36) amide, human - FAQs
View additional product information for Glucagon-Like Peptide-1 (7-36) amide, human - FAQs (J66197.EXD, J66197.MCR)
1 product FAQs found
What is the water solubility of Glucagon-Like Peptide-1 (7-36) amide, human (Cat. No. J66197)?
Glucagon-Like Peptide-1 (7-36) amide, human has a solubility of 1 mg/mL in water.