Bradykinin (2-9)
Bradykinin (2-9)
Bradykinin (2-9)
Thermo Scientific Chemicals

Bradykinin (2-9)

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J66218.MB
also known as J66218-MB
Price (USD)
473.00
Each
Add to cart
Quantity:
25 mg
Request bulk or custom format
Price (USD)
473.00
Each
Add to cart
Specifications
FormatPowder
Molecular Weight (g/mol)904.02
Product TypeBradykinin (2-9)
Purification MethodHPLC
Quantity25 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C