Sulbactam, 98%
Sulbactam, 98%
Sulbactam, 98%
Thermo Scientific Chemicals

Sulbactam, 98%

An antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies. | CAS: 68373-14-8 | C8H11NO5S | 233.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66235.06
also known as J66235-06
Price (USD)
181.65
Special offer
Online exclusive
Ends: 30-Jun-2026
213.00
Save 31.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
181.65
Special offer
Online exclusive
Ends: 30-Jun-2026
213.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS68373-14-8
IUPAC Name(2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular FormulaC8H11NO5S
InChI KeyFKENQMMABCRJMK-RITPCOANSA-N
SMILESCC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥97.5%
FormPowder
Sulbactam is an antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only