Angiotensinogen (1-14), Porcine
Angiotensinogen (1-14), Porcine
Thermo Scientific Chemicals

Angiotensinogen (1-14), Porcine

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J66239.LB0
also known as J66239-#0
5 mg
Catalog number J66239.LB0
also known as J66239-#0
Price (USD)
379.65
Online Exclusive
422.00
Save 42.35 (10%)
Each
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)
379.65
Online Exclusive
422.00
Save 42.35 (10%)
Each
Add to cart
Specifications
FormatLyophilized Solid
Molecular Weight (g/mol)1759.04
Product TypeAngiotensinogen (1-14), porcine
Purification MethodHPLC
Quantity5 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C