D-Fructose 1,6-bisphosphate trisodium salt octahydrate, 98%
D-Fructose 1,6-bisphosphate trisodium salt octahydrate, 98%
D-Fructose 1,6-bisphosphate trisodium salt octahydrate, 98%
Thermo Scientific Chemicals

D-Fructose 1,6-bisphosphate trisodium salt octahydrate, 98%

CAS: 81028-91-3 | C6H26Na3O20P2 | 549.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Catalog number J66256.14
also known as J66256-14
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
116.65
Online Exclusive
129.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS81028-91-3
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥97.5%
Water Content24.0-27.0%
Appearance (Color)White to off-white
D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.

Solubility
Soluble in water

Notes
Moisture Sensitive, store away from water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Lei Lv et. al. Mitogenic and Oncogenic Stimulation of K433 Acetylation Promotes PKM2 Protein Kinase Activity and Nuclear Localization. Molecular Cell, 2013,52(3), 340-352.
  2. Pranav Puri; Anisha Goel; Agnieszka Bochynska; Bert Poolman. Regulation of acetate kinase isozymes and its importance for mixed-acid fermentation in Lactococcus lactis. Journal of Bacteriology, 2014,196(7), 1386-1393.