[Tyr<sup>15</sup>] Fibrinopeptide B, Human
Thermo Scientific Chemicals

[Tyr15] Fibrinopeptide B, Human

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J66258.MA
also known as J66258-MA
10 mg
Catalog number J66258.MA
also known as J66258-MA
Price (USD)
381.65
Special offer
Online exclusive
Ends: 30-Jun-2026
449.00
Save 67.35 (15%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
381.65
Special offer
Online exclusive
Ends: 30-Jun-2026
449.00
Save 67.35 (15%)
Each
Specifications
FormatSolid
Molecular Weight (g/mol)1715.73
Product Type[Tyr15] Fibrinopeptide B, human
Purification MethodHPLC
Quantity10 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C