N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
Thermo Scientific Chemicals

N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

For protein solubilization | CAS: 14933-08-5 | C17H37NO3S | 335.547 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J66265.14
also known as J66265-14
Price (USD)
198.65
Special offer
Online exclusive
Ends: 31-Dec-2025
233.00
Save 34.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 31-Dec-2025
233.00
Save 34.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS14933-08-5
IUPAC Name3-(dodecyldimethylazaniumyl)propane-1-sulfonate
Molecular FormulaC17H37NO3S
InChI KeyIZWSFJTYBVKZNK-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min.
For protein solubilization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only