Amyloid beta (1-40)
Thermo Scientific Chemicals

Amyloid beta (1-40)

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66277.EXD
also known as J66277-!D
Price (USD)
279.65
Online Exclusive
310.00
Save 30.35 (10%)
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
310.00
Save 30.35 (10%)
Each
Specifications
FormatPowder
Molecular Weight (g/mol)4329.79
Product TypeAmyloid beta (1-40)
Purification MethodHPLC
Quantity0.5 mg
View more
Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer’s disease and neurodegeneration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C