Puromycin aminonucleoside, 98%
Puromycin aminonucleoside, 98%
Puromycin aminonucleoside, 98%
Thermo Scientific Chemicals

Puromycin aminonucleoside, 98%

Used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability. | CAS: 58-60-6 | C12H18N6O3 | 294.315 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66301.MC
also known as J66301-MC
Price (USD)
484.65
Special offer
Online exclusive
Ends: 30-Jun-2026
570.00
Save 85.35 (15%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
484.65
Special offer
Online exclusive
Ends: 30-Jun-2026
570.00
Save 85.35 (15%)
Each
Chemical Identifiers
CAS58-60-6
IUPAC Name4-amino-2-[6-(dimethylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolan-3-ol
Molecular FormulaC12H18N6O3
InChI KeyRYSMHWILUNYBFW-UHFFFAOYNA-N
SMILESCN(C)C1=C2N=CN(C3OC(CO)C(N)C3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Solution TestSolubility: Clear and colorless to pale yellow solution at 50 mg/ml concentration in water
FormPowder
Melting Point≥216°C
Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Puromycin aminonucleoside is used as a puromycin aminonucleoside useful in the study of human glomerular disease, and glomerular function and morphology. Used to probe endothelial glycosaminoglycan synthesis in cultured glomerular endothelial cells and their relation to cell permeability.

Solubility
Soluble in water at 50mg/ml. May require heating

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only