Netilmicin sulfate, 595 ^mg/mg
Netilmicin sulfate, 595 ^mg/mg
Netilmicin sulfate, 595 ^mg/mg
Thermo Scientific Chemicals

Netilmicin sulfate, 595 ^mg/mg

A broad spectrum antibiotic | CAS: 56391-57-2 | C21H43N5O11S | 573.659 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
25 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66302.MA
also known as J66302-MA
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Specifications
Boiling Point868°C
CAS56391-57-2
Chemical Name or MaterialNetilmicin sulfate
EINECS Number260-147-3
Formula Weight720.75
View more
Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. William A. Craig.; Sigurdur Gudmundsson.; Richard M. Reich . Netilmicin Sulfate: A Comparative Evaluation of Antimicrobial Activity, Pharmacokinetics, Adverse Reactions and Clinical Efficacy. Pharmacotherapy: The Journal of Human Pharmacology and Drug Therapy. 1983, 3, (6), 305-314.
  2. Adams, E.; Puelings, D.; Rafiee, M.; Roets, E.; Hoogmartens, J. Determination of netilmicin sulfate by liquid chromatography with pulsed electrochemical detection. Journal of Chromatography A. 1998, 812, (1-2), 151-157.