2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%
2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%
2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%
Thermo Scientific Chemicals

2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%

CAS: 14131-84-1 | C12H20O6 | 260.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number J66303.14
also known as J66303-14
Price (USD)
229.65
Online Exclusive
255.00
Save 25.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
229.65
Online Exclusive
255.00
Save 25.35 (10%)
Each
Specifications
Beilstein84382
CAS14131-84-1
Chemical Name or Material2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose
ColorWhite
Formula Weight260.28
View more
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

Solubility
Soluble in acetone, methanol, water.

Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only
Shunya Takahashi; Nobuyuki Hishinuma; Hiroyuki Koshino; Tadashi Nakata. Synthesis of ovalicin starting from D-mannose. Journal of Organic Chemistry. 2005, 70, (4), 10162-10165.