Cefprozil, Thermo Scientific Chemicals
Cefprozil, Thermo Scientific Chemicals
Cefprozil, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cefprozil, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J66326.MC
also known as J66326-MC
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66326.MC
also known as J66326-MC
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
146.00
Save 21.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 28-Dec-2026
146.00
Save 21.35
Each
Specifications
Boiling Point803°C
CAS92665-29-7
Chemical Name or MaterialCefprozil
Flash Point>110°C (230°F)
Formula Weight389.42
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only