Mitoxantrone Dihydrochloride
Mitoxantrone Dihydrochloride
Mitoxantrone Dihydrochloride
Mitoxantrone Dihydrochloride
Thermo Scientific Chemicals

Mitoxantrone Dihydrochloride

CAS: 70476-82-3 | C22H30Cl2N4O6, C22H28N4O6.2HCl | 517.40 g/mol
Have Questions?
Catalog NumberQuantity
J66341.MF
also known as J66341-MF
50 mg
Catalog number J66341.MF
also known as J66341-MF
Price (USD)
162.00
Each
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)
162.00
Each
Add to cart
Chemical Identifiers
CAS70476-82-3
IUPAC Namedihydrogen 1,4-dihydroxy-5,8-bis({2-[(2-hydroxyethyl)amino]ethyl}amino)-9,10-dihydroanthracene-9,10-dione dichloride
Molecular FormulaC22H30Cl2N4O6
InChI KeyZAHQPTJLOCWVPG-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].OCCNCCNC1=CC=C(NCCNCCO)C2=C1C(=O)C1=C(O)C=CC(O)=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to black
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
Assay from Supplier's CofA≥97.0%
Mitoxantrone dihydrochloride is an antiviral, antibacterial, antiprotozoal, immunomodulating, and antineoplastic cytostatic anthraquinone derivative. Induces DNA damage by intercalating into DNA and inhibiting Topo II (topoisomerase II). Mitoxantrone dihydrochloride induces interstrand DNA cross-links and DNA-protein cross-links in cellular systems. Mitoxantrone dihydrochloride has recently been shown to be an inhibitor of DNA methylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only
Contents & Storage
-30°C to -10°C