Mupirocin, 92%
Mupirocin, 92%
Mupirocin, 92%
Thermo Scientific Chemicals

Mupirocin, 92%

CAS: 12650-69-0 | C26H44O9 | 500.629 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66347.MC
also known as J66347-MC
Price (USD)
769.65
Online Exclusive
855.00
Save 85.35 (10%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
769.65
Online Exclusive
855.00
Save 85.35 (10%)
Each
Chemical Identifiers
CAS12650-69-0
IUPAC Name9-{[(2E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-{[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl}oxan-2-yl]-3-methylbut-2-enoyl]oxy}nonanoic acid
Molecular FormulaC26H44O9
InChI KeyMINDHVHHQZYEEK-HBBNESRFSA-N
SMILESC[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]1CO[C@@H](C\C(C)=C\C(=O)OCCCCCCCCC(O)=O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
pH3.5 to 4.5, saturated aqueous solution
Mupirocin is involved in inhibition of protein synthesis was reversed by isoleucine and Pseudomonic Acid was shown to inhibit IleRS (isoleucyl-tRNA synthetase) in vitro. It also demonstrates weak inhibitory action on FARSL (phenylalanyl-tRNA synthetase). Efficacy against staphylococci and streptococci including multiply resistant Staphylcoccus aureus and Staphylococcus epidermidis is reported with Pseudomonic Acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mupirocin is involved in inhibition of protein synthesis was reversed by isoleucine and Pseudomonic Acid was shown to inhibit IleRS (isoleucyl-tRNA synthetase) in vitro. It also demonstrates weak inhibitory action on FARSL (phenylalanyl-tRNA synthetase). Efficacy against staphylococci and streptococci including multiply resistant Staphylcoccus aureus and Staphylococcus epidermidis is reported with Pseudomonic Acid.

Solubility
Soluble in DMSO or methanol. Sparingly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Typical working concentrations are in the range of 0.25µg/mL-0.39µg/mL.
RUO – Research Use Only

General References:

  1. Andrew E Simor; Tammy L Stuart; Lisa Louie; Christine Watt; Marianne Ofner-Agostini; Denise Gravel; Michael Mulvey; Mark Loeb; Allison McGeer; Elizabeth Bryce; Anne Matlow. Mupirocin-resistant, methicillin-resistant Staphylococcus aureus strains in Canadian hospitals. Antimicrobial Agents and Chemotherapy. 2007, 51, (11), 3880-3886.
  2. Gaston H.M. Vondenhoff; Arthur Van Aerschot. Aminoacyl-tRNA synthetase inhibitors as potential antibiotics. European Journal of Medicinal Chemistry. 2011, 46, (11), 5227-5236.