Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals
Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals
Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J66355.03
also known as J66355-03
1 g
Catalog number J66355.03
also known as J66355-03
Price (USD)
63.70
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
63.70
Each
Specifications
CAS23661-28-1
Chemical Name or MaterialPhenyl 2,3,4,6-Tetra-O-acetyl-beta-D-thioglucopyranoside
ColorWhite
Formula Weight440.47
IUPAC Name[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only