Lactobionic acid, 97%
Lactobionic acid, 97%
Lactobionic acid, 97%
Thermo Scientific Chemicals

Lactobionic acid, 97%

CAS: 96-82-2 | C12H22O12 | 358.296 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
Catalog number J66368.14
also known as J66368-14
Price (USD)
78.65
Online Exclusive
86.80
Save 8.15 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
78.65
Online Exclusive
86.80
Save 8.15 (9%)
Each
Chemical Identifiers
CAS96-82-2
IUPAC Name(2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanoic acid
Molecular FormulaC12H22O12
InChI KeyJYTUSYBCFIZPBE-AMTLMPIISA-N
SMILESOC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)5.0% max.
Assay (Titration)97% min.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only