Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
Solubility
Soluble in water (1.19 g/L) at 20°C, and ethanol.
Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only
General References:
- Mao Shen; Wenping Jia; Caiping Lin; Guodong Fan; Yangmin Jin; Xiaoying Chen; Guang Chen. Facile synthesis of folate-conjugated magnetic/fluorescent bifunctional microspheres. Nanoscale Research Letters. 2014, 9, (1), 558.
- Juan C. Domínguez-Romero; Juan F. García-Reyes; Antonio Molina-Díaz. Comparative evaluation of seven different sample treatment approaches for large-scale multiclass sport drug testing in urine by liquid chromatography-mass spectrometry. Journal of Chromatography A. 2013, 1361,