Is the CREBtide peptide acetylated at the N-terminus?No, the CREBtide peptide is not acetylated at the N-terminus.
What is the amino acid sequence of the CREBtide peptide?The amino acid sequence of the CREBtide peptide is: Lys-Arg-Arg-Glu-Ile-Leu-Ser-Arg-Arg-Pro-Ser-Tyr-Arg