Meropenem trihydrate, 98-101%
Meropenem trihydrate, 98-101%
Meropenem trihydrate, 98-101%
Thermo Scientific Chemicals

Meropenem trihydrate, 98-101%

CAS: 119478-56-7 | C17H25N3O5S | 383.46 g/mol
Have Questions?
Catalog NumberQuantity
J66446.MF50 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66446.MF
Price (USD)
198.65
Special offer
Online exclusive
Ends: 28-Dec-2026
234.00
Save 35.35
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 28-Dec-2026
234.00
Save 35.35
Each
Chemical Identifiers
CAS119478-56-7
IUPAC Name(4R)-3-{[(3S,5S)-5-(dimethylcarbamoyl)pyrrolidin-3-yl]sulfanyl}-6-[(1S)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid trihydrate
Molecular FormulaC17H31N3O8S
InChI KeyCTUAQTBUVLKNDJ-TXBRDXQXSA-N
SMILESO.O.O.C[C@H](O)C1C2[C@@H](C)C(S[C@@H]3CN[C@@H](C3)C(=O)N(C)C)=C(N2C1=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Proton NMRConforms to structure
Appearance (Color)White to pale yellow
Assay (unspecified)98.0-101.0%
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.

Solubility
Soluble in aqueous solution at approximately 5mg/ml

Notes
RUO – Research Use Only

General References:

  1. Hiroshi Yamamoto.; Fumio Yamamoto.; Kazuyuki Ishibashi.; Mamika Motokawa. In situ replacement with equine pericardial roll grafts for ruptured infected aneurysms of the abdominal aorta. Journal of Vascular Surgery. 2009, 49, (4), 1041-1045.
  2. Hiromichi Yamaguchi.; Yuko Komase.; Mizuki Ikehara.; Takahito Yamamoto.; Toshihito Shinagawa. Disseminated cryptococcal infection with eosinophilia in a healthy person. Journal of Infection and Chemotherapy. 2008, 14, (4), 319-324.