Angiotensin II (3-8), Human
Angiotensin II (3-8), Human
Angiotensin II (3-8), Human
Thermo Scientific Chemicals

Angiotensin II (3-8), Human

 
Have Questions?
Catalog NumberQuantity
J66457.LB0
also known as J66457-#0
5 mg
Catalog number J66457.LB0
also known as J66457-#0
Price (USD)
70.65
Online Exclusive
77.90
Save 7.25 (9%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
77.90
Save 7.25 (9%)
Each
Chemical Identifiers
CAS23025-68-5
SpecificationsSpecification SheetSpecification Sheet
Molecular WeightConsistent (theoretical MW 774.90)
FormLyophilized powder
Assay from Supplier's CofA≥94.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C