1-Deoxynojirimycin hydrochloride, 98%
1-Deoxynojirimycin hydrochloride, 98%
1-Deoxynojirimycin hydrochloride, 98%
Thermo Scientific Chemicals

1-Deoxynojirimycin hydrochloride, 98%

CAS: 73285-50-4 | C6H14ClNO4 | 199.631 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
25 mg
Catalog number J66458.MB
also known as J66458-MB
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
298.65
Online Exclusive
332.00
Save 33.35 (10%)
Each
Specifications
Beilstein3563225
CAS73285-50-4
Chemical Name or Material1-Deoxynojirimycin hydrochloride
ColorWhite
Formula Weight199.64
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only