Ceftazidime pentahydrate, 95%
Ceftazidime pentahydrate, 95%
Ceftazidime pentahydrate, 95%
Thermo Scientific Chemicals

Ceftazidime pentahydrate, 95%

CAS: 78439-06-2 | C22H32N6O12S2 | 636.648 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66460.03
also known as J66460-03
Price (USD)
132.65
Online exclusive
147.00
Save 14.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
132.65
Online exclusive
147.00
Save 14.35
Each
Chemical Identifiers
CAS78439-06-2
IUPAC Name1-{[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(1-carboxy-1-methylethoxy)imino]acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}-1,2-dihydropyridin-2-ylium pentahydrate
Molecular FormulaC22H32N6O12S2
InChI KeyNMVPEQXCMGEDNH-TZVUEUGBSA-N
SMILESO.O.O.O.O.CC(C)(ON=C(/C(=O)N[C@H]1[C@H]2SCC(CN3[CH+]C=CC=C3)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Loss on Drying13.0-15.0%
Appearance (Color)White to off-white
Assay from Supplier's CofA≥94.0%
Ceftazidime pentahydrate is an antibacterial agent. It is used especially for Pseudomonas and other gram-negative infections in debilitated patients. It is used in the treatment of patients with infections caused by susceptible strains of organisms in the following diseases: lower respiratory tract infections,skin and skin structure infections, urinary tract infections, bacterial septicemia, bone and joint infections, gynecologic infections, intra abdominal infections (including peritonitis), and central nervous system infections (including meningitis).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in aqueous solution

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

General References:

  1. Borys Snopok; Ivanna Kruglenko. Analyte induced water adsorbability in gas phase biosensors: the influence of ethinylestradiol on the water binding protein capacity. Analyst (Cambridge UK).2015, 140, (9), 3225-3232.
  2. A Adu; C L Armour. Drug utilisation review (DUR) of the third generation cephalosporins. Focus on ceftriaxone, ceftazidime and cefotaxime. Drugs.1995, 50, (3), 423-439.