Agarose, Electrophoresis Grade
Agarose, Electrophoresis Grade
Agarose, Electrophoresis Grade
Agarose, Electrophoresis Grade
Thermo Scientific Chemicals

Agarose, Electrophoresis Grade

CAS: 9012-36-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number J66501.18
also known as J66501-18
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.20
Save 13.55 (15%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.20
Save 13.55 (15%)
Each
Add to cart
Chemical Identifiers
CAS9012-36-6
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H38O19
InChI KeyMJQHZNBUODTQTK-WKGBVCLCSA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Melting point: 87 to 89°C
pH5.0 to 6.0 (in gel)
Protease activityNone detected
Appearance (Color)White
Assay (special)Electroendosmosis (EEO): ≤ 0.12%
View more
Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.

Solubility
Soluble in water(10g/L).

Notes
Gel strength (1.5% w/v): >1120g/cm^2; Gelling temperature: 36-39°C; EEO: <0.12
RUO – Research Use Only

General References:

  1. James M. Bailey.; Norman Davidson. Methylmercury as a reversible denaturing agent for agarose gel electrophoresis. Analytical Biochemistry. 1976, 70, (1), 75-85.
  2. Carl-Bertil Laurell. Quantitative estimation of proteins by electrophoresis in agarose gel containing antibodies. Analytical Biochemistry. 1966, 15, (1), 45-52.