Ampicillin trihydrate
Ampicillin trihydrate
Ampicillin trihydrate
Thermo Scientific Chemicals

Ampicillin trihydrate

Commonly used to select for ampicillin resistance in mutated and transformed cells
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66514.14
also known as J66514-14
Price (USD)
94.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
94.00
Each
Chemical Identifiers
CAS7177-48-2
IUPAC Name(2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
Molecular FormulaC16H25N3O7S
InChI KeyRXDALBZNGVATNY-CWLIKTDRSA-N
SMILESO.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)12-15%
Appearance (Color)White
Assay from Supplier's CofA90.0%-105.0% (anhydrous basis)
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
View more
Commonly used to select for ampicillin resistance in mutated and transformed cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
Keep cold