Humanin, human
Thermo Scientific Chemicals

Humanin, human

 
Have Questions?
Catalog NumberQuantity
J66525.LB0
also known as J66525-#0
5 mg
Catalog number J66525.LB0
also known as J66525-#0
Price (USD)
453.65
Online Exclusive
504.00
Save 50.35 (10%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
453.65
Online Exclusive
504.00
Save 50.35 (10%)
Each
Specifications
FormatSolid
Molecular Weight (g/mol)2687.22
Product TypeHumanin, human
Purification MethodHPLC
Quantity5 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C