Cefotiam dihydrochloride, Thermo Scientific Chemicals
Cefotiam dihydrochloride, Thermo Scientific Chemicals
Cefotiam dihydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cefotiam dihydrochloride, Thermo Scientific Chemicals

A broad spectrum antibiotic
Have Questions?
Catalog NumberQuantity
J66529.MC
also known as J66529-MC
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66529.MC
also known as J66529-MC
Price (USD)
368.65
Online Exclusive
409.00
Save 40.35 (10%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
368.65
Online Exclusive
409.00
Save 40.35 (10%)
Each
Specifications
Boiling Point94°C
CAS66309-69-1
Chemical Name or MaterialCefotiam dihydrochloride
EINECS Number266-312-6
Flash Point>110°C (230°F)
View more
A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only