2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, 98%
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, 98%
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, 98%
Thermo Scientific Chemicals

2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose, 98%

CAS: 20880-92-6 | C12H20O6 | 260.286 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
Catalog number J66537.18
also known as J66537-18
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Specifications
CAS20880-92-6
Chemical Name or Material2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose
ColorWhite
Formula Weight260.28
IUPAC Name[(3aR,5aS,8aS)-2,2,7,7-tetramethyl-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[4,5-a
View more
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose is used as chiral auxiliaries in Michael and Aldol addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose is used as chiral auxiliaries in Michael and Aldol addition reactions.

Solubility
Soluble in chloroform, ethyl acetate, methanol.

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
RUO – Research Use Only

General References:

  1. Pinheiro S. ; Pedraza S. F. ; Peralta M. A. ; Carvalho E. M. ; Farias F. M. C. ; Ferreira V. F. 1,2:5,6-Di-O-Isopropylidene-Alpha -D-Glucofuranose and 2,3 : 4,5-Di-O-Isopropylidene-Beta -D-Fructopyranose as Chiral Auxiliaries in Michael and Aldol Addition Reactions. Journal of carbohydrate chemistry.1998, 17, 901-913.
  2. T.Lis and A.Weichsel. Structure of 2,3:4,5-di-O-isopropylidene-[beta]-D-fructopyranose. Acta Cryst.1987, C43, 1954-1956.