Cecropin A, Porcine
Thermo Scientific Chemicals

Cecropin A, Porcine

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J66546.EXD
also known as J66546-!D
Price (USD)
265.65
Online Exclusive
295.00
Save 29.35 (10%)
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
265.65
Online Exclusive
295.00
Save 29.35 (10%)
Each
Specifications
FormatLyophilized Powder
Molecular Weight (g/mol)4003.77
Product TypeCecropin A, porcine
Purification MethodHPLC
Quantity0.5 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C