2-Keto-D-gluconic acid hemicalcium salt monohydrate, 99%
2-Keto-D-gluconic acid hemicalcium salt monohydrate, 99%
2-Keto-D-gluconic acid hemicalcium salt monohydrate, 99%
Thermo Scientific Chemicals

2-Keto-D-gluconic acid hemicalcium salt monohydrate, 99%

CAS: 3470-37-9 | C12H20CaO14 | 428.356 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number J66609.06
also known as J66609-06
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Specifications
Beilstein3808734
CAS3470-37-9
Chemical Name or Material2-Keto-D-gluconic acid hemicalcium salt monohydrate
ColorWhite
Formula Weight231.22
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only