Adrenomedullin (13-52), Human
Thermo Scientific Chemicals

Adrenomedullin (13-52), Human

 
Have Questions?
Catalog NumberQuantity
J66619.MCR
also known as J66619-M^
1 mg
Catalog number J66619.MCR
also known as J66619-M^
Price (USD)
511.65
Special offer
Online exclusive
Ends: 28-Dec-2026
602.00
Save 90.35
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
511.65
Special offer
Online exclusive
Ends: 28-Dec-2026
602.00
Save 90.35
Each
Specifications
FormatSolid
Molecular Weight (g/mol)4533.06
Product TypeAdrenomedullin (13-52), human
Purification MethodHPLC
Quantity1 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C